WHA68761304
Whatman® GD/X syringe filters
polysulfone membrane, pore size 0.45 μm, diam. 13 mm, non-sterile, pk of 50 ea
Synonym(s):
Whatman filter, Z286575, syringe filter, syringe filters
Select a Size
$770.00
Select a Size
About This Item
$770.00
Recommended Products
material
polypropylene housing
polysulfone membrane
sterility
non-sterile
feature
glass fiber prefilter
packaging
pk of 50 ea
manufacturer/tradename
Whatman 6876-1304
Whatman Article No. 28420516 (US reference)
diam.
13 mm
volume
≤10 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Related Categories
General description
Features and Benefits
- Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)