WHA111156
Whatman® polycarbonate Nuclepore filter discs 0.1 μm pore size
black, hydrophilic, track-etched, DEFT, 100 ea, 47 mm diam
Synonym(s):
Z759589, nuclepore membrane, polycarbonate membrane
About This Item
Recommended Products
product name
Whatman® Nuclepore™ Track-Etched Membranes, diam. 47 mm, pore size 0.2 μm, polycarbonate, black, pkg of 100 ea
material
polycarbonate
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 111156
Whatman Article No. 28417652 (US reference)
diam.
47 mm
color
black
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
General description
Black membranes for rapid standard plate count results
Cytiva′s Nuclepore membrane filters are compatible with the direct epifluorescent microscopy filter technique (DEFT), which allows microorganisms to be counted directly on a filter. DEFT can be conducted in under half an hour, sidestepping the standard > 24 hour process of incubating a microbial sample. Cytiva′s irgalan black membranes greatly reduce background fluorescence, improving visibility and helping to distinguish microorganisms from other particulates.
Microscopic membrane filters
Cytiva′s microscopic membrane filters are made by track-etching – the polycarbonate is bombarded with radiation and chemically treated to create sharp, uniform filter pores. The PCTE membrane is designed to trap microbes efficiently, and can be more effective than other filter media for certain microorganisms.
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Features and Benefits
- Smooth, gridded PCTE membrane surface efficiently supports microorganism enumeration in a standard plate count
- Irgalan black membranes for reduced background fluorescence in support of epifluorescence microscopy
- Extremely low nonspecific absorption
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service