Key Documents
WHA10441003
Whatman® glass vacuum filtration device
GV025 series, GV025/0/03 glass frit, diam. 25 mm
Synonym(s):
Z750751
Select a Size
1 PKG
$99.30
$99.30
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$99.30
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
$99.30
Please contact Customer Service for Availability
material
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
packaging
pk of 1
manufacturer/tradename
Whatman 10441003
Whatman Article No. 28415543 (US reference)
diam.
25 mm
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
General description
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
This product is only the "glass frit".
Features and Benefits
  • Accessory to GV025 Glass Filter Funnel
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)