WHA10400014
Whatman® nitrocellulose filter discs 12 μm pore size
white, hydrophilic, 100 ea, 50 mm diam
Synonym(s):
Whatman filter, Z696927, cellulose membrane, membrane filter
Select a Size
$381.00
Select a Size
About This Item
$381.00
Recommended Products
Product Name
Whatman® nitrocellulose membrane filters, pore size 12 μm, diam. 50 mm, pack of 100 ea
sterility
non-sterile
feature
hydrophilic
packaging
pack of 100 ea
manufacturer/tradename
Whatman 10400014
Whatman Article No. 28414921 (US reference)
diam.
50 mm
pore size
12 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Narrow pore size distribution for improved surface capture and analysis.
- Low levels of extractables to ensure sample integrity.
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service