Key Documents
WHA10370381
Whatman® glass microfiber filters with organic binder, Grade GF 10
roll, W × L 30 mm × 20 m, pack of 1
Synonym(s):
fiber filters, glass filter, whatman filter
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
material
glass membrane (with organic binder)
roll
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Whatman 10370381
Whatman Article No. 28414797 (US reference)
W × L
30 mm × 20 m
General description
This filter with extreme mechanical stability and temperature resistant up to 180°C is used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)