Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W779547

Sigma-Aldrich

γ-Cyclodextrin

Wacker Chemie AG, ≥98% (on dry basis)

Synonym(s):

Cavamax® W8, Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
Beilstein:
78740
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reg. compliance

FDA 21 CFR 117

description

derived from GMO corn

Assay

≥98% (on dry basis)

manufacturer/tradename

Wacker Chemie AG

impurities

≤0.5% reducing substances (determined as dextrose)
≤1.000 microorganisms per g
≤11% water
≤20 ppm volatile organics
≤5 ppm heavy metals
0 salmonella / E. Coli in 10 g

ign. residue

≤0.1%

cation traces

As: ≤1.3 ppm
Pb: ≤1 ppm

SMILES string

[H][C@]1(O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O)O[C@@H]2[C@@H](COC(C)=O)O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O

InChI

1S/C48H80O40/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-72H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-/m1/s1

InChI key

GDSRMADSINPKSL-HSEONFRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

γ-Cyclodextrin (γ-CD), a water-soluble cyclic oligosaccharide consisting of eight α-(1,4)-linked glucopyranose subunits, can be used in a wide range of applications including:
  • Synthesis of renewable and biocompatible metal-organic frameworks.[1]
  • Formation of inclusion complexes with a variety of guest molecules to increase their solubility in water and other polar solvents.[2][3]
  • Synthesis of γ-CD based nanogels and dendrimers as carriers and stabilizers for drug delivery applications.[4][5]

Legal Information

Cavamax is a registered trademark of Wacker Chemie AG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclodextrin-mediated enhancement of riboflavin solubility and corneal permeability.
Morrison, Peter WJ et al.
Angewandte Chemie (International Edition in English), 10(2), 756-762 (2013)
Metal-organic frameworks from edible natural products.
Smaldone, Ronald A et al.
Angewandte Chemie (International Edition in English), 49(46), 8630-8634 (2010)
Cross-linked hydroxypropyl-β-cyclodextrin and γ-cyclodextrin nanogels for drug delivery: physicochemical and loading/release properties.
Moya-Ortega, Maria D et al.
Carbohydrate Polymers, 87(3), 2344-2351 (2012)
γ-Cyclodextrin: a review on enzymatic production and applications.
Li, Zhaofeng et al.
Applied Microbiology and Biotechnology, 77(2), 245-245 (2007)
Cyclodextrin functionalized graphene nanosheets with high supramolecular recognition capability: synthesis and host- guest inclusion for enhanced electrochemical performance.
Guo, Yujing et al.
ACS Nano, 4(7), 4001-4010 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service