Skip to Content
MilliporeSigma
All Photos(1)

Documents

W6756

2-Ketoglutaric acid-1-13C

API for Clinical Studies, ≥99 atom % 13C

Synonym(s):

α-Ketoglutaric acid-1-13C, 2-Ketopentanedioic acid-1-13C, 2-Oxoglutaric acid-1-13C, 2-Oxopentanedioic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH2CO13CO2H
CAS Number:
Molecular Weight:
147.09
MDL number:
UNSPSC Code:
12352115

grade

cGMP

isotopic purity

≥99 atom % 13C

mp

114-117 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

OC(=O)CCC(=O)[13C](O)=O

InChI

1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)/i5+1

InChI key

KPGXRSRHYNQIFN-HOSYLAQJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service