Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W6373

Sigma-Aldrich

Potassium palmitate-1,2,3,4-13C4

API for Clinical Studies, 99 atom % 13C

Synonym(s):

Hexadecanoic acid-1,2,3,4-13C4 potassium salt, Palmitic acid-1,2,3,4-13C4 potassium salt, Potassium hexadecanoate-1,2,3,4-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(13CH2)313CO2K
CAS Number:
Molecular Weight:
298.49
MDL number:
UNSPSC Code:
12352001

grade

cGMP

isotopic purity

99 atom % 13C

mp

262-263 °C
285 °C (dec.) (lit.)

mass shift

M+4

storage temp.

15-25°C

SMILES string

[K+].CCCCCCCCCCCC[13CH2][13CH2][13CH2][13C]([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i13+1,14+1,15+1,16+1;

InChI key

MQOCIYICOGDBSG-OZGJQOAZSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service