Skip to Content
MilliporeSigma
All Photos(1)

Documents

W530659

Sigma-Aldrich

Tobacco extract

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12164502

biological source

(nicotiana tabacum)

grade

FG
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

refractive index

n20/D 1.508

density

0.973 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

tobacco

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service