Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530179

Sigma-Aldrich

Macadamia oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502

biological source

Macadamia integrifolia

grade

Certified organic (NOP)
Halal
Kosher

origin

Kenya origin

refractive index

n20/D 1.468

density

0.840 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

Organoleptic

nutty

Legal Information

Certified organic by MOSA

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

410.0 °F

Flash Point(C)

210 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service