Skip to Content
MilliporeSigma
All Photos(1)

Documents

W523704

Sigma-Aldrich

Rose oil,Turkish

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12164502

biological source

Rosa spp.

origin

France origin

refractive index

n20/D 1.4570

density

0.861 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

oily; rose

Features and Benefits

A fragrance material extracted from "Mon" rose flowers grown in northeastern Thailand utilizing a state-of-the-art technology wherby oils are prepared at ambient temperature and without the use of high vacuum, alcohol or hexane. The clear dark red mobile liquid possesses the aroma properties of fresh rose blossom. Mon Rose Oil is insoluble in water, and soluble in ethanol and vegetable oils.

Preparation Note

Extraction method: steam distillation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 82-82 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 41-41 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service