Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W521604

Sigma-Aldrich

Cornmint oil

natural, chinese type, FG

Synonym(s):

Mentha arvensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
4219
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Mentha arvensis

Quality Level

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor pressure

0.1 mmHg ( 20 °C)

origin

China origin

refractive index

n20/D 1.460 (lit.)

bp

218 °C/760 mmHg (lit.)
218.4 °C/755 mmHg

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

minty; camphoraceous

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1.0%
Taste at 10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.0 °F - closed cup

Flash Point(C)

65.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 53-53 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service