Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W519308

Sigma-Aldrich

3-Methyl-3-buten-1-ol

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1071239
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.176
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥97%

refractive index

n20/D 1.433 (lit.)

bp

130-132 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity

SMILES string

CC(=C)CCO

InChI

1S/C5H10O/c1-5(2)3-4-6/h6H,1,3-4H2,2H3

InChI key

CPJRRXSHAYUTGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-3-buten-1-ol is a volatile aliphatic alcohol found in the extracts of basil and thyme leaves, immature mangaba fruits and some honey samples.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components of mangaba fruit (Hancornia speciosa Gomes) at three stages of maturity
Sampaio TS and Nogueira PCL
Food Chemistry, 95(4), 606-610 (2006)
Identification of volatile components in basil (Ocimum basilicum L.) and thyme leaves (Thymus vulgaris L.) and their antioxidant properties.
Lee SJ, et al.
Food Chemistry, 91(1), 131-137 (2005)
[Penetration of primary and tertiary alcohol isomers through rubber and isolated skin].
A L Mikhaleva et al.
Gigiena truda i professional'nye zabolevaniia, (11)(11), 46-47 (1982-01-01)
Rosa A Pérez et al.
Journal of agricultural and food chemistry, 50(9), 2633-2637 (2002-04-18)
Headspace solid-phase microextraction (SPME), followed by gas chromatography (GC)-mass spectrometry (MS) determination, has been used for the analysis of honey volatiles. Two SPME fibers were employed to study the composition of volatiles from various types of Spanish honeys. The best

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service