Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W519200

Sigma-Aldrich

Isophytol

>95%

Synonym(s):

3,7,11,15-Tetramethyl-1-hexadecen-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40O
CAS Number:
Molecular Weight:
296.53
Beilstein:
1783243
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.168

biological source

synthetic

grade

Kosher

Assay

>95%

refractive index

n20/D 1.457 (lit.)

bp

309 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

chocolate

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C=C

InChI

1S/C20H40O/c1-7-20(6,21)16-10-15-19(5)14-9-13-18(4)12-8-11-17(2)3/h7,17-19,21H,1,8-16H2,2-6H3

InChI key

KEVYVLWNCKMXJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

343.4 °F - closed cup

Flash Point(C)

173 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service