Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W518123

Sigma-Aldrich

2-Methyl-2-thiazoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NS
CAS Number:
Molecular Weight:
101.17
Beilstein:
104274
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.086

biological source

synthetic

Assay

98%

color

colorless

refractive index

n20/D 1.520 (lit.)

bp

144-145 °F
144.0-145.0 °C (lit.)

mp

-101.0 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)
1.067 g/mL at 25 °C

application(s)

flavors and fragrances

SMILES string

CC1=NCCS1

InChI

1S/C4H7NS/c1-4-5-2-3-6-4/h2-3H2,1H3

InChI key

JUIQOABNSLTJSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Schaal et al.
Comptes rendus de l'Academie des sciences. Serie III, Sciences de la vie, 313(7), 319-325 (1991-01-01)
Intranasal injections of a fragrant solution of 2-methyl-2-thiazoline elicited significant heart rate decelerations in late pregnancy fetal sheep, while the injection of a control fluid (isotonic saline) had no effect. This result indicates that the ovine fetuses are able to
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (10)(10), 55-56 (1980-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service