Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W512508

Sigma-Aldrich

3,7-Dimethyl-2,6-octadienenitrile

mixture of isomers, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCN
CAS Number:
Molecular Weight:
149.23
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.05

biological source

synthetic

Assay

≥97%

refractive index

n20/D 1.475 (lit.)

bp

110 °C/1.3 mmHg (lit.)

density

0.853 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

C\C(C)=C\CC\C(C)=C\C#N

InChI

1S/C10H15N/c1-9(2)5-4-6-10(3)7-8-11/h5,7H,4,6H2,1-3H3/b10-7+

InChI key

HLCSDJLATUNSSI-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetyana Kobets et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 115, 228-243 (2018-03-20)
Genotoxicity of flavor and fragrance materials was assessed in Turkey Egg Genotoxicity Assay (TEGA) using 32P-nucleotide postlabeling (NPL) and comet assays to detect hepatic DNA adducts and strand breaks. Twenty materials having results in GADD45a-Gluc 'BlueScreen HC' genotoxicity assay, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service