Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W510866

Sigma-Aldrich

2-Propyl-4-methyl-1,3-dioxolane, mixture of isomers

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.095

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.412 (lit.)

bp

137 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CCCC1OCC(C)O1

InChI

1S/C7H14O2/c1-3-4-7-8-5-6(2)9-7/h6-7H,3-5H2,1-2H3

InChI key

WRTYJFJOOTWPJP-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service