Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W510262

Sigma-Aldrich

3-(4-Methoxyphenyl)-1-propanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4(CH2)3OH
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
Flavis number:
2.173

biological source

synthetic

Assay

99%

form

viscous liquid

color

colorless

refractive index

n20/D 1.532 (lit.)

bp

164-168 °C/18 mmHg (lit.)
164-168 °F/18 mmHg

mp

26 °C (lit.)

Organoleptic

vegetable

SMILES string

COc1ccc(CCCO)cc1

InChI

1S/C10H14O2/c1-12-10-6-4-9(5-7-10)3-2-8-11/h4-7,11H,2-3,8H2,1H3

InChI key

NIIDHUCLROLCBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service