Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W507318

Sigma-Aldrich

4-tert-Butylcyclohexyl acetate

≥98%

Synonym(s):

4-tert-Butylcyclohexyl acetate, mixture of cis and trans, woody acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H10C(CH3)3
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.452 (lit.)

bp

228-230 °C/25 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody

SMILES string

CC(=O)OC1CCC(CC1)C(C)(C)C

InChI

1S/C12H22O2/c1-9(13)14-11-7-5-10(6-8-11)12(2,3)4/h10-11H,5-8H2,1-4H3

InChI key

MBZRJSQZCBXRGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service