Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W503002

Sigma-Aldrich

1,2,3-Trimethoxybenzene

≥97%

Synonym(s):

Pyrogallol trimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OCH3)3
CAS Number:
Molecular Weight:
168.19
Beilstein:
1910422
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
4.084

Assay

≥97%

bp

241 °C (lit.)

mp

43-47 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(OC)c1OC

InChI

1S/C9H12O3/c1-10-7-5-4-6-8(11-2)9(7)12-3/h4-6H,1-3H3

InChI key

CRUILBNAQILVHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service