Skip to Content
MilliporeSigma
All Photos(1)

Documents

W502901

Sigma-Aldrich

2-Methylfuran

≥98%

Synonym(s):

Silvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
Beilstein:
103733
EC Number:
Council of Europe no.:
2209
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.030

biological source

synthetic

grade

Kosher

vapor density

2.8 (vs air)

vapor pressure

139 mmHg ( 20 °C)
561 mmHg ( 55 °C)

Assay

≥98%

contains

≤300 ppm BHT as stabilizer

refractive index

n20/D 1.433 (lit.)

bp

63-66 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

Organoleptic

ethereal

SMILES string

Cc1ccco1

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

VQKFNUFAXTZWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiao Huo et al.
Mutation research, 854-855, 503209-503209 (2020-07-15)
We have measured the toxicity and genotoxicity of 2-methylfuran, which is formed in foods during thermal processing. The agent was administered by oral gavage to male Sprague-Dawley rats, daily for 28 days, before performing general toxicology analysis and the following
Luca Cappelletti et al.
Ultrasonics sonochemistry, 27, 30-36 (2015-07-19)
An ultrasound-assisted three-component, one-pot domino reaction with ferrocenecarboxaldehyde is herein reported. The sequence of reactions entails the allylindation and dehydrative alkylation of stabilized C-nucleophiles (e.g. electron-rich-(hetero)aromatics and stabilized enols) and N-nucleophiles (e.g. azoles). Sonochemical reactions have been performed in three
Hanna Valolahti et al.
Global change biology, 21(9), 3478-3488 (2015-05-23)
Emissions of biogenic volatile organic compounds (BVOCs) have been earlier shown to be highly temperature sensitive in subarctic ecosystems. As these ecosystems experience rapidly advancing pronounced climate warming, we aimed to investigate how warming affects the BVOC emissions in the
Maryam Chaichi et al.
Food additives & contaminants. Part B, Surveillance, 8(1), 73-80 (2014-10-31)
In this study, the levels of furan, 2-methylfuran, 2,5-dimethylfuran, vinyl furan, 2-methoxymethyl-furan and furfural in different coffee products were evaluated. Simultaneous determination of these six furanic compounds was performed by a head space liquid-phase micro-extraction (HS-LPME) method. A total of
Benjamin Sachse et al.
Carcinogenesis, 35(10), 2339-2345 (2014-07-24)
Furfuryl alcohol is a rodent carcinogen present in numerous foodstuffs. Sulfotransferases (SULTs) convert furfuryl alcohol into the DNA reactive and mutagenic 2-sulfoxymethylfuran. Sensitive techniques for the isotope-dilution ultra performance liquid chromatography-tandem mass spectrometry quantification of resulting DNA adducts, e.g. N

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service