Skip to Content
MilliporeSigma
All Photos(1)

Documents

W474505

Sigma-Aldrich

6-Methoxy-2,6-dimethylheptanal

≥95%, stabilized

Synonym(s):

Methoxymelonal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
FEMA Number:
4745
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; floral; citrus

storage temp.

2-8°C

InChI

1S/C10H20O2/c1-9(8-11)6-5-7-10(2,3)12-4/h8-9H,5-7H2,1-4H3

InChI key

QMXBURPYNWBMJR-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service