Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W473401

Sigma-Aldrich

3-(Methylthio)decanal

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22OS
CAS Number:
Molecular Weight:
202.36
FEMA Number:
4734
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.306
NACRES:
NA.21

Quality Level

Assay

≥90%

form

liquid

refractive index

n20/D 1.475

density

0.930 g/mL at 25 °C

application(s)

flavors and fragrances

Organoleptic

fatty; green; meaty; vegetable; roasted; sulfurous

storage temp.

2-8°C

SMILES string

[H]C(CC(SC)CCCCCCC)=O

InChI

1S/C11H22OS/c1-3-4-5-6-7-8-11(13-2)9-10-12/h10-11H,3-9H2,1-2H3

InChI key

WZRMFIROSHXUJB-UHFFFAOYSA-N

Application

(1) Potential Applications: This compound imparts an impacting sulfurous green vegetative fattiness that would be useful in broccoli, cabbage and other vegetables and fruits such as cucumber and watermelon. It also has a warm roasted fatty chicken depth nuance.
(2) Odor Characteristics: @1.0% ETOH. Sulfurous and fatty. Green vegetative, broccoli and cabbage, fatty with notes of roasted chicken.
(3) Taste Characteristics: @ 0.20 PPM. Impacting sulfurous green vegetative with notes of cucumber and watermelon with fatty nuances of cooked egg.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service