Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W441200

Sigma-Aldrich

cis-3-Nonen-1-ol

95%, stabilized

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
142.24
FEMA Number:
4412
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.234
NACRES:
NA.21

biological source

synthetic

Assay

95%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.45 (lit.)

bp

107-109 °C/20 mmHg (lit.)

density

0.843 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; melon; waxy

storage temp.

2-8°C

SMILES string

CCCCC\C=C/CCO

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h6-7,10H,2-5,8-9H2,1H3/b7-6-

InChI key

IFTBJDZSLBRRMC-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service