Skip to Content
MilliporeSigma
All Photos(1)

Documents

W438933

Sigma-Aldrich

2,4-Dimethylpyridine

≥98%, FG

Synonym(s):

2,4-Lutidine, 2,4-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
FEMA Number:
4389
Beilstein:
1506
MDL number:
UNSPSC Code:
12352100
Flavis number:
14.104
NACRES:
NA.21

grade

FG

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU 1223/2009
EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.499 (lit.)

bp

159 °C (lit.)

mp

−60 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

smoky; phenolic

SMILES string

Cc1ccnc(C)c1

InChI

1S/C7H9N/c1-6-3-4-8-7(2)5-6/h3-5H,1-2H3

InChI key

JYYNAJVZFGKDEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service