Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W431600

Sigma-Aldrich

2′-Methylacetophenone

≥98%

Synonym(s):

1-(2-Methylphenyl)ethanone, 1-(2-Tolyl)ethanone, 2-Acetyltoluene, o-Methylacetophenone, Methyl o-tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
FEMA Number:
4316
Beilstein:
907005
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.251

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.5318 (lit.)

bp

214 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

almond; cherry

SMILES string

CC(=O)c1ccccc1C

InChI

1S/C9H10O/c1-7-5-3-4-6-9(7)8(2)10/h3-6H,1-2H3

InChI key

YXWWHNCQZBVZPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible applications: Vanilla, cherry, almond, coumarin and various nut nuances.

Biochem/physiol Actions

Odor at 1.0%: Sweet, naphthyl, cherry and almond, solvent-like phenol and toluene-like with berry, floral and coumarin-like nuances.
Taste at 5ppm: Nutty, cherry, almond, and coumarinic-like.

Other Notes

Natural occurrence: Carambola.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerard Mosciano
Perfumer & Flavorist, 34 (1), 58-59 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service