Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W428600

Sigma-Aldrich

2-Isobutyl-4-methyl-1,3-dioxolane

mixture of isomers, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
FEMA Number:
4286
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.135
NACRES:
NA.21

biological source

synthetic

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.414 (lit.)

bp

150-153 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; ethereal; buttery; sweet

SMILES string

CC(C)CC1OCC(C)O1

InChI

1S/C8H16O2/c1-6(2)4-8-9-5-7(3)10-8/h6-8H,4-5H2,1-3H3

InChI key

PDVLTWPJDBXATJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible applications: Italian cheeses, dairy applications, apple, pineapple and cocoa nuances.

Biochem/physiol Actions

Odor at 10.0%: Sweet, ethereal, musty and slightly fermented, dairy-like with fruity apple and banana nuances.
Taste at 20ppm: Creamy, cocoa, woody, juicy fruit with apply, pineapple and dairy cheesy nuances.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerard Mosciano
Perfumer & Flavorist, 34 (1), 58-59 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service