Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W424001

Sigma-Aldrich

(±)-sec-Butylamine

99%

Synonym(s):

(±)-2-Aminobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(NH2)CH3
CAS Number:
Molecular Weight:
73.14
FEMA Number:
4240
Beilstein:
1361345
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.005

biological source

synthetic

Assay

99%

refractive index

n20/D 1.3928 (lit.)

bp

63 °C (lit.)

mp

−72 °C (lit.)

density

0.724 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

fishy

SMILES string

CCC(C)N

InChI

1S/C4H11N/c1-3-4(2)5/h4H,3,5H2,1-2H3

InChI key

BHRZNVHARXXAHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service