All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2NH2
CAS Number:
Molecular Weight:
59.11
FEMA Number:
4237
Beilstein:
1098243
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.004
biological source
synthetic
vapor density
2 (vs air)
vapor pressure
4.79 psi ( 20 °C)
Assay
≥99%
autoignition temp.
604 °F
expl. lim.
10.4 %
refractive index
n20/D 1.388 (lit.)
bp
48 °C (lit.)
mp
−83 °C (lit.)
density
0.719 g/mL at 25 °C (lit.)
SMILES string
CCCN
InChI
1S/C3H9N/c1-2-3-4/h2-4H2,1H3
InChI key
WGYKZJWCGVVSQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
<-31.0 °F
Flash Point(C)
< -35 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ryan R Gordon et al.
PloS one, 9(9), e104271-e104271 (2014-09-10)
To identify molecular alterations in prostate cancers associating with relapse following neoadjuvant chemotherapy and radical prostatectomy patients with high-risk localized prostate cancer were enrolled into a phase I-II clinical trial of neoadjuvant chemotherapy with docetaxel and mitoxantrone followed by prostatectomy.
Sebastian Finger et al.
Biological chemistry, 395(7-8), 769-778 (2014-07-09)
The binding of cationic polyamines to negatively charged lipid membranes is driven by electrostatic interactions and additional hydrophobic contributions. We investigated the effect of polyamines with different number of charges and charge separation on the phase transition behavior of vesicles
Claire Bagnéris et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8428-8433 (2014-05-23)
Voltage-gated sodium channels are important targets for the development of pharmaceutical drugs, because mutations in different human sodium channel isoforms have causal relationships with a range of neurological and cardiovascular diseases. In this study, functional electrophysiological studies show that the
Yuki Itakura et al.
PloS one, 10(9), e0136660-e0136660 (2015-09-04)
Rhythmic motor patterns underlying many types of locomotion are thought to be produced by central pattern generators (CPGs). Our knowledge of how CPG networks generate motor patterns in complex nervous systems remains incomplete, despite decades of work in a variety
Lionel Faure et al.
Phytochemistry, 110, 58-71 (2014-12-11)
Alkamides and N-acylethanolamines (NAEs) are bioactive, amide-linked lipids that influence plant development. Alkamides are restricted to several families of higher plants and some fungi, whereas NAEs are widespread signaling molecules in both plants and animals. Fatty acid amide hydrolase (FAAH)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service