Skip to Content
MilliporeSigma
All Photos(1)

Documents

W420201

Sigma-Aldrich

Prenyl acetate

≥98%, stabilized, FG

Synonym(s):

3,3-dimethyl allyl acetate, 3-methyl-2-buten-1-ol acetate, 3-methyl-2-buten-1-yl acetate, 3-methyl-2-butenyl acetate, Isopent-2-enyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
128.17
FEMA Number:
4202
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.692
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.43 (lit.)

bp

151-152 °C/752 mmHg (lit.)

solubility

H2O: insoluble
alcohol: soluble

density

0.917 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; fruity

SMILES string

CC(=O)OC\C=C(\C)C

InChI

1S/C7H12O2/c1-6(2)4-5-9-7(3)8/h4H,5H2,1-3H3

InChI key

XXIKYCPRDXIMQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric M Jones et al.
Cell systems, 8(3), 254-260 (2019-03-25)
G protein-coupled receptors (GPCRs) are central to how mammalian cells sense and respond to chemicals. Mammalian olfactory receptors (ORs), the largest family of GPCRs, mediate the sense of smell through activation by small molecules, though for most bonafide ligands, they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service