Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W401900

Sigma-Aldrich

Farnesal, mixture of isomers

≥85%

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrienal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
FEMA Number:
4019
Beilstein:
1723427
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.148
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Assay

≥85%

refractive index

n20/D >1.4920 (lit.)

bp

126-129 °C/3.5 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

minty

SMILES string

[H]C(=O)\C=C(/C)CC\C=C(/C)CC\C=C(/C)C

InChI

1S/C15H24O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11-12H,5-6,8,10H2,1-4H3/b14-9+,15-11+

InChI key

YHRUHBBTQZKMEX-YFVJMOTDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grace Jones et al.
The FEBS journal, 273(21), 4983-4996 (2006-10-27)
The in vivo ligand-binding function and ligand-binding activity of the Drosophila melanogaster retinoid-X receptor (RXR) ortholog, ultraspiracle, toward natural farnesoid products of the ring gland were assessed. Using an equilibrium fluorescence-binding assay, farnesoid products in the juvenile hormone (JH) biosynthesis
H Inoue et al.
Biochemical and biophysical research communications, 196(3), 1401-1405 (1993-11-15)
Farnesal and 3-hydroxy-2,3-dihydrofarnesal (3-hydroxy-3,7,11-trimethyl-6,10-dodecadiene-1-al) were formed from farnesol when the alcohol was incubated with the protoplast of Botryococcus braunii B race strain. This fact suggests the existence of farnesal hydratase in the alga. Feeding experiments showed that both farnesal and
Kenji Shimomura et al.
Journal of chemical ecology, 36(8), 824-833 (2010-07-08)
Callosobruchus rhodesianus (Pic) (Coleoptera: Chrysomelidae: Bruchinae) is a pest of stored legumes through the Afro-tropical region. In laboratory bioassays, males of C. rhodesianus were attracted to volatiles collected from virgin females. Collections were purified by various chromatographic techniques, and the
Constantinos Tsangarakis et al.
The Journal of organic chemistry, 73(7), 2905-2908 (2008-03-07)
The sesquiterpene nanaimoal was synthesized in 21% overall yield and in a biomimetic manner. As a key step, the acid-catalyzed cyclization of farnesal under zeolite NaY confinement conditions was used. The intrazeolite cyclization of farnesal affords as major product a
Anna Luxová et al.
Chirality, 16(4), 228-233 (2004-03-23)
The absolute configurations of citronellol, 2,3-dihydrofarnesol, and 2,3-dihydrofarnesal in male marking pheromones of seven species of bumblebees and cuckoo bumblebees were determined by enantioselective gas chromatography on a capillary column coated with 60% heptakis(2,3-di-O-acetyl-6-O-TBDMS)-beta-cyclodextrin in polysiloxane PS 268. Pure (-)-S-enantiomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service