Skip to Content
MilliporeSigma
All Photos(1)

Documents

W398608

Sigma-Aldrich

4-Hydroxybenzoic acid

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
FEMA Number:
3986
Beilstein:
970950
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Flavis number:
8.040
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥99%

mp

213-217 °C (lit.)

solubility

methanol: 5%, clear to slightly hazy, colorless to faintly yellow

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

phenolic

SMILES string

OC(=O)c1ccc(O)cc1

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

FJKROLUGYXJWQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Letian X Xie et al.
The Journal of biological chemistry, 287(28), 23571-23581 (2012-05-18)
Most of the Coq proteins involved in coenzyme Q (ubiquinone or Q) biosynthesis are interdependent within a multiprotein complex in the yeast Saccharomyces cerevisiae. Lack of only one Coq polypeptide, as in Δcoq strains, results in the degradation of several
L C Goudas et al.
Anesthesia and analgesia, 89(5), 1209-1215 (1999-11-30)
Intracerebroventricular (ICV) morphine administration is effective for the management of refractory cancer pain. Recent preclinical observations of acute depletion of the major endogenous intracellular antioxidant glutathione (GSH) in brain and peripheral organs after ICV morphine in rodents led us to
L I Wiebe et al.
Drug metabolism and disposition: the biological fate of chemicals, 6(3), 296-302 (1978-05-01)
Butylated hydroxytoluene (BHT) containing the stable isotope 13C was synthesized from 2-[13C]methylpropan-2-ol. A minor constituent of urine following ingestion of BHT-13C by a human volunteer was identified as 3,5-di-(1-[13C]methyl-1-methylethyl)-4-hydroxybenzoic acid, The major metabolite detected was 13C-labeled 5-carboxy-7-(1-carboxy-1-methylethyl)-3,3-dimethyl-2-hydroxy-2,3-dihydrobenzofuran. Detailed spectral analysis
Suri Roowi et al.
Molecular nutrition & food research, 53 Suppl 1, S68-S75 (2009-05-06)
Human urine was collected over a 24 h period after the consumption of 250 mL of (i) water, (ii) orange juice, and (iii) orange juice plus 150 mL of full fat natural yoghurt. The orange juice contained 168 micromol of
Mireia Urpi-Sarda et al.
Analytical and bioanalytical chemistry, 394(6), 1545-1556 (2009-04-01)
Proanthocyanidins, flavonoids exhibiting cardiovascular protection, constitute a major fraction of the flavonoid ingested in the human diet. Although they are poorly absorbed, they are metabolized by the intestinal microbiota into various phenolic acids. An analytical method, based on an optimized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service