Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

W395500

Sigma-Aldrich

3-Nonen-2-one solution

10% in ethanol, natural (US), FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O
CAS Number:
Molecular Weight:
140.22
FEMA Number:
3955
Council of Europe no.:
11163
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.188
grade:
FG
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 872/2012

concentration

10% in ethanol

refractive index

n20/D 1.370

density

0.810 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; mushroom; musty; spicy; vegetable; nutty

SMILES string

CCCCC\C=C\C(C)=O

InChI

1S/C9H16O/c1-3-4-5-6-7-8-9(2)10/h7-8H,3-6H2,1-2H3/b8-7+

InChI key

HDKLIZDXVUCLHQ-BQYQJAHWSA-N

General description

Natural occurence(neat): Asparagus, chicken, crayfish, mushroom, oats, soybeans, apricot and beef

Application

The powerful earthy, fatty notes of this material will enhance mushroom, vegetable flavors like cucumber and cultured dairy flavors like sour cream, yogurt and kefir. It should also be considered to enlarge the ketone complex in Brie, Gorgonzola, Camembert and other mold-ripened cheese flavors.

Biochem/physiol Actions

Odor at 10%
Taste at 1-5ppm

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(2), 49-49 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service