Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W394512

Sigma-Aldrich

p-Anisic acid

natural, 99%, FG

Synonym(s):

4-Methoxybenzoic acid, p-Methoxybenzoic acid, Draconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3945
Beilstein:
508910
EC Number:
Council of Europe no.:
10077
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.071
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

99%

mp

182-185 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

animal

SMILES string

COc1ccc(cc1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ZEYHEAKUIGZSGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Anisic acid is one of the key constituents of star-anise.[1][2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic and magnetic properties of Cu (II) complexes with selected biologically important ligands.
Swislocka R, et al.
open chemistry, 10(4), 1095-1105 (2012)
Extraction and Analysis on Water-soluble Volatile Constituents from Star-anise.
Wang T-Y
Journal of Anhui Agricultural Sciences, 14, 004-004 (2009)
N Dodoff et al.
Journal of inorganic biochemistry, 60(4), 257-266 (1995-12-01)
The complexes [Pt(NH3)(pmbah)Cl2], [Pt(NH3)(pcbah)Cl2], [Pt(pmbah)2X2] and [Pt(pcbah)2X2] (pmbah = 4-methoxybenzoic acid hydrazide, pcbah = 4-chlorobenzoic acid hydrazide; X = Cl, Br, I) have been synthesized and characterized by elemental analysis, electric conductivity, 1H NMR, IR, and electronic spectra. A cis-square
Stephen G Bell et al.
Dalton transactions (Cambridge, England : 2003), 41(28), 8703-8714 (2012-06-15)
The crystal structures of the 4-methoxybenzoate bound forms of cytochrome P450 enzymes CYP199A2 and CYP199A4 from the Rhodopseudomonas palustris strains CGA009 and HaA2 have been solved. The structures of these two enzymes, which share 86% sequence identity, are very similar
A Hage et al.
Applied microbiology and biotechnology, 52(6), 834-838 (2000-01-05)
Ligninolytic basidiomycetes were screened for their ability to reduce aryl acids to the corresponding aldehydes and alcohols. Seven fungal strains converted p-anisic acid in high molar yields to the reduced products. The white-rot fungus Bjerkandera sp. strain BOS55 was one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service