Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W393118

Sigma-Aldrich

cis-3-Hexenyl tiglate

≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
182.26
FEMA Number:
3931
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.559

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.46 (lit.)

bp

105 °C/5 mmHg (lit.)

density

0.951 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; earthy; vegetable; herbaceous

SMILES string

CC\C=C/CCOC(=O)\C(C)=C\C

InChI

1S/C11H18O2/c1-4-6-7-8-9-13-11(12)10(3)5-2/h5-7H,4,8-9H2,1-3H3/b7-6-,10-5+

InChI key

JNWQKXUWZWKUAY-JQEGGOPCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1%
Taste at 1-5 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Cao et al.
Insects, 11(7) (2020-07-08)
Thrips hawaiiensis is a common thrips pest of various plant flowers with host preference. Plant volatiles provide important information for host-searching in insects. We examined the behavioral responses of T. hawaiiensis adults to the floral volatiles of Gardenia jasminoides Ellis
Mosciano, G.
Perfumer & Flavorist, 25, 78-78 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service