Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W389201

Sigma-Aldrich

Phenylpyruvic acid

98%

Synonym(s):

2-Oxo-3-phenylpropanoic acid, 2-Oxo-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COCOOH
CAS Number:
Molecular Weight:
164.16
FEMA Number:
3892
Beilstein:
2207312
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
8.109

biological source

synthetic

Assay

98%

mp

150-154 °C (lit.)
158-160 °C

SMILES string

OC(=O)C(=O)Cc1ccccc1

InChI

1S/C9H8O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,11,12)

InChI key

BTNMPGBKDVTSJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Tan et al.
Applied biochemistry and biotechnology, 136(3), 279-289 (2007-07-13)
The high-density fermentation of recombinant Pichia pastoris was carried out in a 1-L fermentor. After 60 h of fermentation, the activities of D-amino acid oxidase (DAAO) and catalase assayed with the permeabilized cells attained 12,532 and 684,800 U/L, respectively. Additionally
Paddy Jim Baggot et al.
Fetal diagnosis and therapy, 23(3), 245-248 (2008-04-18)
Organic acids were examined from normal and Down syndrome pregnancies to identify possible differences between the amniotic fluid from fetuses with Down Syndrome compared with that of normal fetuses. Amniotic fluids were obtained from prior amniocenteses. Forty-one normal and 22
Valeriano Dal Cin et al.
The Plant cell, 23(7), 2738-2753 (2011-07-14)
Altering expression of transcription factors can be an effective means to coordinately modulate entire metabolic pathways in plants. It can also provide useful information concerning the identities of genes that constitute metabolic networks. Here, we used ectopic expression of a
Gerrit J Poelarends et al.
Biochemistry, 46(33), 9596-9604 (2007-07-31)
The enzymatic conversion of cis- or trans-3-chloroacrylic acid to malonate semialdehyde is a key step in the bacterial degradation of the nematocide 1,3-dichloropropene. Two mechanisms have been proposed for the isomer-specific hydrolytic dehalogenases, cis- and trans-3-chloroacrylic acid dehalogenase (cis-CaaD and
Tapan Kanti Paine et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(21), 6073-6081 (2007-05-01)
Iron(II)-phenylpyruvate complexes of tetradentate tris(6-methyl-2-pyridylmethyl)amine (6-Me3-TPA) and tridentate benzyl bis(2-quinolinylmethyl)amine (Bn-BQA) were prepared to gain insight into C-C bond cleavage catalyzed by dioxygenase enzymes. The complexes we have prepared and characterized are [Fe(6-Me3-tpa)(prv)][BPh4] (1), [Fe2(6-Me3-tpa)2(pp)][(BPh4)2] (2), and [Fe2(6-Me3-tpa)2(2'-NO2-pp)][(BPh4)2] (3), [Fe(6-Me3-tpa)(pp-Me)][BPh4]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service