Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W382418

Sigma-Aldrich

1-Decen-3-ol

≥98%

Synonym(s):

3-Hydroxy-1-decene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH(OH)CH=CH2
CAS Number:
Molecular Weight:
156.27
FEMA Number:
3824
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.136

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.444 (lit.)

bp

214-217 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cheese; green; waxy; earthy; fruity

SMILES string

CCCCCCCC(O)C=C

InChI

1S/C10H20O/c1-3-5-6-7-8-9-10(11)4-2/h4,10-11H,2-3,5-9H2,1H3

InChI key

NSFWLHKFTGZFBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1.0% EtOH
Taste at 1-10 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.0 °F - closed cup

Flash Point(C)

91.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 49-49 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service