Skip to Content
MilliporeSigma
All Photos(1)

Documents

W378621

Sigma-Aldrich

γ-Methyl decalactone

96%, FG

Synonym(s):

γ-Methyl-γ-decalactone, 4-Methyl-4-decanolide, 5-Hexyldihydro-5-methyl-2(3H)-furanone, Dihydrojasmone lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3786
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.051
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

96%

refractive index

n20/D 1.450

density

0.942 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; fatty; oily; cheesy; waxy

SMILES string

O=C(CC1)OC1(CCCCCC)C

InChI

1S/C11H20O2/c1-3-4-5-6-8-11(2)9-7-10(12)13-11/h3-9H2,1-2H3

InChI key

ALWUKGXLBSQSMA-UHFFFAOYSA-N

Application

The profile of this product will enhance dairy flavor profiles where cream plays a major role, namely milk, butter, cream, ice cream, crème brulée, cream cheese, cheese, etc. It can enlarge the fatty complex and add richness in coconut and nuts like pecan macadamia, cashew and almond, as well as in meat fat flavors such as tallow, lard and schmaltz.

Biochem/physiol Actions

Odor at 10%
Taste at 5-10ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 50-50 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service