Skip to Content
MilliporeSigma
All Photos(1)

Documents

W377500

Sigma-Aldrich

cis-1-(1-Ethoxyethoxy)-3-hexene

mixture of isomers, natural (US), 95%, FG

Synonym(s):

Acetaldehyde ethyl cis-3-hexenyl acetal, Ethyl cis-3-hexenyl acetal acetaldehyde, Leaf acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3775
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.081
NACRES:
NA.21

grade

FG
Fragrance grade
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

95%

refractive index

n20/D 1.422

density

0.850 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; leafy; mushroom; musty; vegetable; violet

InChI

1S/C10H20O2/c1-4-6-7-8-9-12-10(3)11-5-2/h6-7,10H,4-5,8-9H2,1-3H3/b7-6-

InChI key

PAEBAEDUARAOSG-SREVYHEPSA-N

General description

Natural occurence: Guava, plum, strawberry

Application

The powerful, green earthy notes of this product will enhance vegetable flavors like green pepper, green beans, asparagus, cucumber, celery, lettuce, radish, watercress, broccoli and bean sprouts. These same notes, at much lower levels, can also add an unripe note to fruits, if such a quality is needed for the profile.

Biochem/physiol Actions

Odor at 1%
Taste at 2-5ppm

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.0 °F - closed cup

Flash Point(C)

50.56 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 38(10), 60-60 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service