Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W376177

Sigma-Aldrich

5-Methyl-2-hepten-4-one

greener alternative

natural (US), ≥97%, mixture of isomers, FG

Synonym(s):

Filbertone, Hazelnut ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$326.00
25 G
$577.00
100 G
$1,780.00

$326.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
5 G
$326.00
25 G
$577.00
100 G
$1,780.00

About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3761
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.139
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural (US)
biological source:
botanical
food allergen:
no known allergens

$326.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥97%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

hazelnut; nutty

SMILES string

O=C(C(CC)C)\C=C\C

InChI

1S/C8H14O/c1-4-6-8(9)7(3)5-2/h4,6-7H,5H2,1-3H3/b6-4+

InChI key

ARJWAURHQDJJAC-GQCTYLIASA-N

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service