Skip to Content
MilliporeSigma
All Photos(1)

Documents

W374806

Sigma-Aldrich

L-Menthyl lactate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O3
CAS Number:
Molecular Weight:
228.33
FEMA Number:
3748
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.551

biological source

synthetic

Assay

≥97%

optical activity

[α]20/D −81°, c = 5 in ethanol

bp

142 °C/5 mmHg (lit.)

mp

42-47 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

alcohol; minty

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@H](C)O

InChI

1S/C13H24O3/c1-8(2)11-6-5-9(3)7-12(11)16-13(15)10(4)14/h8-12,14H,5-7H2,1-4H3/t9-,10?,11+,12-/m1/s1

InChI key

UJNOLBSYLSYIBM-SGUBAKSOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1.0%
Taste at 25 ppm

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 56-56 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 41-41 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service