Skip to Content
MilliporeSigma
All Photos(1)

Documents

W372404

Sigma-Aldrich

2-Pentadecanone

≥98%, FG

Synonym(s):

Methyl tridecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO(CH2)12CH3
CAS Number:
Molecular Weight:
226.40
FEMA Number:
3724
Beilstein:
1764872
EC Number:
Council of Europe no.:
11808
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.137
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

bp

293 °C (lit.)

mp

37-41 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; spicy

SMILES string

CCCCCCCCCCCCCC(C)=O

InChI

1S/C15H30O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(2)16/h3-14H2,1-2H3

InChI key

CJPNOLIZCWDHJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eriko Harada et al.
PloS one, 7(1), e29710-e29710 (2012-01-13)
Odorant-binding proteins (OBPs) are extracellular proteins found in insect chemosensilla, where they participate in the sensing of odors, tastes, and pheromones. Although a large number of OBP genes have been identified in insect genomes, their molecular functions and biological roles
George F Antonious et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 50(9), 667-673 (2015-06-17)
Glandular trichomes on the leaves of wild tomato, Lycopersicon hirsutum f. hirsutum Mull, also known as Solanum habrochaites (Solanaceae) synthesize and accumulate high levels of methyl ketones (MKs). The potential of using MKs as alternatives to synthetic acaricides for controlling
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service