Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W358207

Sigma-Aldrich

1-Octen-3-yl acetate

≥97%, FCC, FG

Synonym(s):

3-Acetoxy octene, Amy vinyl carbinol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3582
EC Number:
Council of Europe no.:
11716
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.281
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.425 (lit.)

bp

80 °C/15 mmHg (lit.)

density

0.878 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lavender; herbaceous

SMILES string

CCCCCC(OC(C)=O)C=C

InChI

1S/C10H18O2/c1-4-6-7-8-10(5-2)12-9(3)11/h5,10H,2,4,6-8H2,1,3H3

InChI key

DOJDQRFOTHOBEK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.0 °F - closed cup

Flash Point(C)

73.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service