Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W356511

Sigma-Aldrich

D-Dihydrocarvone, mixture of isomers

natural, 97%, FG

Synonym(s):

p-Menth-8-en-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
3565
EC Number:
Council of Europe no.:
11703
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.128
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

97%

refractive index

n20/D 1.472 (lit.)

bp

87-88 °C/6 mmHg (lit.)

density

0.926 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caraway; spicy; herbaceous; ethereal; woody; minty

SMILES string

CC1CCC(CC1=O)C(C)=C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-9H,1,4-6H2,2-3H3

InChI key

AZOCECCLWFDTAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurence: Caraway seed, celery seed, dill, patchouli, spearmint, buchu and pepper

Application

The dual, herbal-minty quality of this compound will add an interesting twist to mint flavors as well as to celery, caraway, dill and herbal flavors, especially intended for cough drops and oral care products. At very low levels, it will add depth to tropical flavors like lychee, guava, passion fruit and rambutan.[1]

Biochem/physiol Actions

Odor at 1%[1]
Taste at 2-4ppm[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 52-52 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service