Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W351318

Sigma-Aldrich

1,9-Nonanedithiol

≥95%, FG

Synonym(s):

1,9-Dimercaptononane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)9SH
CAS Number:
Molecular Weight:
192.39
FEMA Number:
3513
EC Number:
Council of Europe no.:
11558
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.069
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥95%

refractive index

n20/D 1.4999 (lit.)

bp

284 °C (lit.)

density

0.952 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

SCCCCCCCCCS

InChI

1S/C9H20S2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2

InChI key

GJRCLMJHPWCJEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible uses: mushroom, meat, chicken, cereal, cucumber.

Biochem/physiol Actions

Odor at 0.10%: waxy, sulfurous, metallic, vegetative.
Taste @ 1 ppm: waxy, mushroom, vegetative with cucumber-like nuances.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, Gerard
Perfumer & Flavorist, 33, 73-73 (2008)
Wei Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 11(35), 4509-4516 (2015-06-04)
The ability to tune gauge factors in terms of magnitude and orientation is important for wearable and conformal electronics. Herein, a sensor device is described which is fabricated by assembling and printing molecularly linked thin films of gold nanoparticles on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service