Skip to Content
MilliporeSigma
All Photos(1)

Documents

W346918

Sigma-Aldrich

Propiophenone

≥99%

Synonym(s):

Ethyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC2H5
CAS Number:
Molecular Weight:
134.18
FEMA Number:
3469
Beilstein:
606215
EC Number:
Council of Europe no.:
599
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.040
NACRES:
NA.21

biological source

synthetic

vapor pressure

1 mmHg ( 50 °C)

Assay

≥99%

refractive index

n20/D 1.526 (lit.)

bp

218 °C (lit.)

mp

17-19 °C (lit.)

density

1.009 g/mL at 20 °C
1.009 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

Organoleptic

floral; fruity; sweet

SMILES string

CCC(=O)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

KRIOVPPHQSLHCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Tang et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 21(3), 236-241 (2015-02-07)
Nontuberculous mycobacteria infection is a growing global concern, but data from Asia are limited. This study aimed to describe the distribution and antibiotic susceptibility profiles of rapidly growing mycobacterium (RGM) isolates in Singapore. Clinical RGM isolates with antibiotic susceptibility tests
Ruben De Pauw et al.
Journal of chromatography. A, 1420, 129-134 (2015-10-20)
A new system is proposed for applying fast temperature gradients in liquid chromatography. It consists of a 0.7 mm × 150 mm fused-silica column coated with a 50 μm Nickel-layer, which is connecting with a power source and a temperature
Zsolt Székelyhidi et al.
Bioorganic & medicinal chemistry letters, 15(13), 3241-3246 (2005-06-01)
SR protein-specific kinase-1 (SRPK-1) has been identified as a validated target for hepatitis B virus (HBV). A series of novel tricyclic quinoxaline derivatives was designed and synthesised as potential kinase inhibitory antiviral agents and was found to be active and
Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak
D Essaid et al.
Journal of pharmaceutical and biomedical analysis, 114, 227-240 (2015-06-24)
Porphyrins are a class of photosensitizers used in photodynamic therapy (PDT). Understanding the interaction of porphyrins with membrane cells components is important in order to improve this therapy. Many analytical methods can be used for this aim. High performance liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service