Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W341304

Sigma-Aldrich

2-Methylpentanal

≥97%

Synonym(s):

2-Methylvaleraldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CHO
CAS Number:
Molecular Weight:
100.16
FEMA Number:
3413
Beilstein:
1739423
EC Number:
Council of Europe no.:
706
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.069

biological source

synthetic

vapor density

3.45 (vs air)

Assay

≥97%

autoignition temp.

390 °F

refractive index

n20/D 1.401 (lit.)

bp

119-120 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

ethereal; earthy; fruity

SMILES string

CCCC(C)C=O

InChI

1S/C6H12O/c1-3-4-6(2)5-7/h5-6H,3-4H2,1-2H3

InChI key

FTZILAQGHINQQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service