Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W340200

Sigma-Aldrich

cis-3-Hexenyl butyrate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3402
EC Number:
Council of Europe no.:
2166
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.270
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥98%

refractive index

n20/D 1.431 (lit.)

bp

96 °C/20 mmHg (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; fruity; vegetable

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)CCC

InChI

1S/C10H18O2/c1-3-5-6-7-9-12-10(11)8-4-2/h5-6H,3-4,7-9H2,1-2H3/b6-5-

InChI key

ZCHOPXVYTWUHDS-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaoyao Peng et al.
Food research international (Ottawa, Ont.), 129, 108873-108873 (2020-02-11)
The phenolic compounds and aroma active compounds in feijoa (Acca sellowiana (O.Berg) Burret) juice from four New Zealand grown cultivars (Apollo, Unique, Opal Star, and Wiki Tu) were investigated. A high total phenolic content (maximum 1.89 mg GAE/mL juice) and significant
Lorenzo Boggia et al.
BMC plant biology, 15, 102-102 (2015-04-19)
The dynamics of plant volatile (PV) emission, and the relationship between damaged area and biosynthesis of bioactive molecules in plant-insect interactions, remain open questions. Direct Contact-Sorptive Tape Extraction (DC-STE) is a sorption sampling technique employing non adhesive polydimethylsiloxane tapes, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service