Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W339709

Sigma-Aldrich

Furfuryl pentanoate

≥97%, FG

Synonym(s):

2-Furfuryl pentanoate, Furfuryl valerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
FEMA Number:
3397
EC Number:
Council of Europe no.:
10647
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.068
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥97%

refractive index

n20/D 1.46 (lit.)

bp

82-83 °C/1 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green

SMILES string

CCCCC(=O)OCc1ccco1

InChI

1S/C10H14O3/c1-2-3-6-10(11)13-8-9-5-4-7-12-9/h4-5,7H,2-3,6,8H2,1H3

InChI key

HRYAVTBTUKVHBU-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service