Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W339105

Sigma-Aldrich

3-Acetyl-2,5-dimethylfuran

≥99%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
FEMA Number:
3391
EC Number:
Council of Europe no.:
10921
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.066
NACRES:
NA.21
To inquire about this product W339105, please contact your local MilliporeSigma office or dealer. Contact Technical Service

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥99%

refractive index

n20/D 1.485 (lit.)

bp

62 °C/0.25 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; nutty

SMILES string

CC(=O)c1cc(C)oc1C

InChI

1S/C8H10O2/c1-5-4-8(6(2)9)7(3)10-5/h4H,1-3H3

InChI key

KBSVBCHYXYXDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service